LIF372152444 Life Chemicals Inc.

Thiomorpholin-3-one

  • MDL number MFCD00152870
  • Cas Number : 20196-21-8
  • Molecular Weight : 117.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart