LIF372152464 Life Chemicals Inc.

5-[3-(Dimethylamino)propyl]-1,3,5-triazinane-2-thione

  • MDL number MFCD00522194
  • Cas Number : 304700-48-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart