LIF372152470 Life Chemicals Inc.

4-(4-Thioxo-1,3,5-triazinan-1-yl)butanoic acid

  • MDL number MFCD00522223
  • Cas Number : 247109-16-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart