LIF372152479 Life Chemicals Inc.

1,3-Diazaspiro[4.4]nonane-2,4-dione

  • MDL number MFCD00022403
  • Cas Number : 699-51-4
  • Molecular Weight : 154.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart