LIF372152480 Life Chemicals Inc.

5-Chloro-7-[(4-methylpiperidin-1-yl)methyl]quinolin-8-ol

  • MDL number MFCD01205322
  • Cas Number : 294675-78-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart