LIF372152481 Life Chemicals Inc.

(Diphenyl-phosphinoyl)-acetonitrile

  • MDL number MFCD00444258
  • Cas Number : 23040-22-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart