LIF372152483 Life Chemicals Inc.

3-bromo-4-hydroxy-1lambda6-thiolane-1,1-dione

  • MDL number MFCD00047101
  • Cas Number : 35330-57-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart