LIF372152489 Life Chemicals Inc.

3-hydrazinyl-6-methyl-4H-1,2,4-triazin-5-one

  • MDL number MFCD00662639
  • Cas Number : 38736-23-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart