LIF372152490 Life Chemicals Inc.

4-amino-6-methyl-3-(methylthio)-1,2,4-triazin-5(4H)-one

  • MDL number MFCD02336234
  • Cas Number : 18826-96-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart