LIF372152496 Life Chemicals Inc.

N-[1-Amino-1-phenyl-meth-(Z)-ylidene]-benzenesulfonamide

  • MDL number MFCD00475019
  • Cas Number : 1123883-53-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart