LIF372152497 Life Chemicals Inc.

3-(1,1-Dioxido-3-oxo-1,2-benzisothiazol-2(3H)-yl)propanoic acid

  • MDL number MFCD00758127
  • Cas Number : 83747-21-1
  • Molecular Weight : 255.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart