LIF372152508 Life Chemicals Inc.

1-(Phenylsulfonyl)cyclopropanecarboxylic acid

  • MDL number MFCD00547146
  • Cas Number : 97383-41-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart