LIF372152516 Life Chemicals Inc.

6,7-dimethoxy-1-(4-methoxyphenyl)-1,2,3,4-tetrahydroisoquinoline

  • MDL number MFCD00409941
  • Cas Number : 55580-65-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart