LIF372152518 Life Chemicals Inc.

2-(methylthio)benzo[d]thiazol-6-amine

  • MDL number MFCD00086478
  • Cas Number : 25706-29-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart