LIF372152519 Life Chemicals Inc.

6-Acetyl-2-amino-4,5,6,7-tetrahydro-thieno[2,3-c]pyridine-3-carboxylic acid ethyl ester

  • MDL number MFCD01750487
  • Cas Number : 24237-43-2
  • Molecular Weight : 268.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart