LIF372152520 Life Chemicals Inc.

2-[(3-Morpholin-4-ylpropyl)amino]-5-nitrobenzonitrile

  • MDL number MFCD02939908
  • Cas Number : 451459-94-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart