LIF372152531 Life Chemicals Inc.

1-phenyl-1H-1,2,4-triazole-3,5-diamine

  • MDL number MFCD00522516
  • Cas Number : 14575-59-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart