LIF372152538 Life Chemicals Inc.

1-(tert-butoxycarbonyl)proline

  • MDL number MFCD00520915
  • Cas Number : 59433-50-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart