LIF372152540 Life Chemicals Inc.

(6-methyl-2,4-dioxo-1,2,3,4-tetrahydropyrimidin-5-yl)acetic acid

  • MDL number MFCD03006091
  • Cas Number : 38580-22-2
  • Molecular Weight : 184.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart