LIF372152543 Life Chemicals Inc.

N,6-dimethyl-2-morpholin-4-ylpyrimidin-4-amine

  • MDL number MFCD02998764
  • Cas Number : 381716-92-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart