LIF372152576 Life Chemicals Inc.

1,3-Dioxo-2-phenyl-2,3-dihydro-1H-isoindole-5-carboxylic acid

  • MDL number MFCD00427538
  • Cas Number : 4649-27-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart