LIF372152586 Life Chemicals Inc.

1-benzyl-1H-benzimidazole-2-sulfonic acid

  • MDL number MFCD00426869
  • Cas Number : 90331-20-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart