LIF372152591 Life Chemicals Inc.

4,7,7-Trimethyl-2,3-dioxo-bicyclo[2.2.1]heptane-1-carboxylic acid

  • MDL number MFCD03011711
  • Cas Number : 69842-14-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart