LIF372152592 Life Chemicals Inc.

3,3-Dimethyl-2-oxo-bicyclo[2.2.1]heptane-1-carboxylic acid

  • MDL number MFCD03011710
  • Cas Number : 469-74-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart