LIF372152595 Life Chemicals Inc.

1-Ethyl-2-oxo-1,2-dihydrobenzo[cd]indole-6-sulfonyl chloride

  • MDL number MFCD02656057
  • Cas Number : 32103-15-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart