LIF372152599 Life Chemicals Inc.

9-Methyl-4-oxo-2-pyrrolidin-1-yl-4H-pyrido[1,2-a]pyrimidine-3-carbaldehyde

  • MDL number MFCD01072122
  • Cas Number : 433318-45-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart