LIF372152622 Life Chemicals Inc.

4-[(5Z)-5-(4-Methoxybenzylidene)-2,4-dioxo-1,3-thiazolidin-3-yl]butanoic acid

  • MDL number MFCD02328140
  • Cas Number : 314260-18-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 88
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart