LIF372152626 Life Chemicals Inc.

4-Imino-thiazolidin-2-one

  • MDL number MFCD00154755
  • Cas Number : 19967-65-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart