LIF372152631 Life Chemicals Inc.

1-Isopropyl-1H-benzoimidazole-2-thiol

  • MDL number MFCD02180803
  • Cas Number : 2416-65-1
  • Molecular Weight : 192.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart