LIF372152633 Life Chemicals Inc.

1-[(5-Chloro-2-thienyl)sulfonyl]piperidine-3-carboxylic acid

  • MDL number MFCD02944037
  • Cas Number : 384847-77-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart