LIF372152634 Life Chemicals Inc.

1-[(5-Chloro-2-thienyl)sulfonyl]piperidine-2-carboxylic acid

  • MDL number MFCD02944015
  • Cas Number : 1008038-62-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart