LIF372152652 Life Chemicals Inc.

1-(4-Chloro-phenyl)-4-hydroxy-1H-pyrazole-3-carboxylic acid ethyl ester

  • MDL number MFCD02361221
  • Cas Number : 26502-56-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart