LIF372152659 Life Chemicals Inc.

3-phenylquinoline-2,4-dicarboxylic acid

  • MDL number MFCD03028436
  • Cas Number : 19585-90-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart