LIF372152665 Life Chemicals Inc.

1-(4-Chlorobenzyl)-1H-benzimidazole-2-thiol

  • MDL number MFCD03670986
  • Cas Number : 100622-93-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart