LIF372152678 Life Chemicals Inc.

3-[(E)-Hydroxyimino]-4,7,7-trimethyl-bicyclo[2.2.1]heptane-1-carboxylic acid

  • MDL number MFCD03659904
  • Cas Number : 100055-50-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart