LIF372152684 Life Chemicals Inc.

2-amino-3a,6a-dimethylhexahydro-1,4-methanocyclopenta[c]pyrrole-1(2H)-carboxamide

  • MDL number MFCD03731244
  • Cas Number : 1212089-56-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart