LIF372152686 Life Chemicals Inc.

2-Benzothiazol-2-yl-nicotinic acid

  • MDL number MFCD02670938
  • Cas Number : 728-66-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart