LIF372152702 Life Chemicals Inc.

2,3-Dihydrothiophene-3-ol 1,1-dioxide

  • MDL number MFCD00519588
  • Cas Number : 6211-59-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart