LIF372152704 Life Chemicals Inc.

3-Chloro-tetrahydro-thiophene 1,1-dioxide

  • MDL number MFCD00154885
  • Cas Number : 3844-04-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart