LIF372152706 Life Chemicals Inc.

3-sulfanyl-1lambda6-thiolane-1,1-dione

  • MDL number MFCD00519568
  • Cas Number : 52513-18-5
  • Molecular Weight : 152.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart