LIF372152714 Life Chemicals Inc.

3-Isocyanato-2,3-dihydrothiophene 1,1-dioxide

  • MDL number MFCD03037886
  • Cas Number : 61308-98-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart