LIF372152715 Life Chemicals Inc.

3-isothiocyanato-1lambda6-thiolane-1,1-dione

  • MDL number MFCD00456661
  • Cas Number : 85109-44-0
  • Molecular Weight : 177.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart