LIF372152717 Life Chemicals Inc.

1,1-Dioxidotetrahydro-3-thienyl chloridocarbonate

  • MDL number MFCD00456631
  • Cas Number : 77691-76-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart