LIF372152720 Life Chemicals Inc.

3-(Isothiocyanatomethyl)tetrahydrothiophene 1,1-dioxide

  • MDL number MFCD00456662
  • Cas Number : 85109-45-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart