LIF372152734 Life Chemicals Inc.

Benzo[h][1,6]naphthyridine-5-carbaldehyde

  • MDL number MFCD03085770
  • Cas Number : 69164-27-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart