LIF372152735 Life Chemicals Inc.

2-Chloro-9H-purine

  • MDL number MFCD00137826
  • Cas Number : 1681-15-8
  • Molecular Weight : 154.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart