LIF372152742 Life Chemicals Inc.

3-Bromo-2-nitropyridine

  • MDL number MFCD00955614
  • Cas Number : 54231-33-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart