LIF372152755 Life Chemicals Inc.

4-Chloro-1-(2,4-dimethylphenyl)-1H-pyrazolo[3,4-d]pyrimidine

  • MDL number MFCD04971631
  • Cas Number : 610277-86-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart