LIF372152763 Life Chemicals Inc.

5-(2,4-Dimethoxyphenyl)-1,3,4-thiadiazol-2-amine

  • MDL number MFCD01925389
  • Cas Number : 299932-63-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart