LIF372152785 Life Chemicals Inc.

6-chloro-5-nitropyrimidin-4-amine

  • MDL number MFCD00127772
  • Cas Number : 4316-94-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart